Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNAS Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521702

Catalog No. PIPA521702


Add to cart

Description

Description

PA5-21702 targets XLalphas in IHC (P) and WB applications and shows reactivity with Human and mouse samples. The PA5-21702 immunogen is a recombit fragment corresponding to a region within amino acids 716 and 998 of GNAS (Uniprot IDNo.Q5JWF2). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

GNAS
Polyclonal
Unconjugated
GNAS
Liquid
GNAS
Recombit fragment corresponding to a region within amino acids 716 and 998 of GNAS (Uniprot IDNo.Q5JWF2)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.78 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O95467, P63094
RP4-543J19.4, AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, PHP1A, PHP1B, PHP1C, POH, adenylate cyclase-stimulating G alpha protein, alternative gene product encoded by XL-exon, extra large alphas protein, guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide 1, guanine nucleotide regulatory protein, guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas, neuroendocrine secretory protein, protein ALEX, secretogranin VI
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
14683, 2778
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit