Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-XLF, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP145933

Catalog No. NBP145933


Add to cart

Description

Description

XLF Polyclonal antibody specifically detects XLF in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

XLF
Unconjugated
PBS, 0.5% BSA, 30% Glycerol with 0.01% Thimerosal
Affinity Purified
NHEJ1
Synthetic peptide of human XLF (NP_079058.1).
Affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Immunoprecipitation, Western Blot
Western Blot 1:500 - 1:2500, Immunoprecipitation
NP_079058.1
Cernunnos, non-homologous end-joining factor 1, nonhomologous end-joining factor 1, Protein cernunnos, XLFFLJ12610, XRCC4-like factor
Rabbit
IgG
0.1 ml
Cancer, Chromatin Research, DNA Repair
Primary
79840
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only