Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPB Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522254

Catalog No. PIPA522254


Add to cart

Description

Description

PA5-22254 targets XPB in ICC, IF, and WB applications and shows reactivity with Human samples. The PA5-22254 immunogen is a recombit fragment corresponding to a region within amino acids 521 and 742 of XPB (Uniprot IDNo.P19447). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

XPB
Polyclonal
Unconjugated
ERCC3
Liquid
ERCC3
Recombit fragment corresponding to a region within amino acids 521 and 742 of XPB (Uniprot IDNo.P19447)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
0.69 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P19447, P49135
BTF2, GTF2H, RAD25, TFIIH, XPB, BTF2 p89, DNA excision repair protein ERCC-3, DNA repair protein complementing XP-B cells, TFIIH 89 kDa subunit, TFIIH basal transcription factor complex 89 kDa subunit, TFIIH basal transcription factor complex helicase XPB subunit, TFIIH p89, basic transcription factor 2 89 kDa subunit, xeroderma pigmentosum group B complementing, xeroderma pigmentosum group B-complementing protein, xeroderma pigmentosum, complementation group B
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
13872, 2071
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit