Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPOT Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514639

Catalog No. PIPA514639

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

XPOT is a protein belonging to the RAN-GTPase exportin family that mediates export of tRNA from the nucleus to the cytoplasm. Translocation of tRNA to the cytoplasm occurs once exportin has bound both tRNA and GTP-bound RAN.
Specifications

Specifications

XPOT
Polyclonal
PBS with 0.09% sodium azide
O43592
Exportin-T; Exportin-T; tRNA exportin; Exportin(tRNA) ; XPOT
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11260
Immunohistochemistry, Western Blot
Unconjugated
XPOT
Liquid
XPOT
KLH conjugated synthetic peptide between 137-166 amino acids from the N-terminal region of human XPOT
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit