Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XRCC3 Rabbit anti-Human, Drosophila, Hamster, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
$175.82 - $436.00
Specifications
Antigen | XRCC3 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:5000, Immunoprecipitation, ICC/IF (Negative) |
Applications | Western Blot, Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100165SS
|
Novus Biologicals
NB100165SS |
0.025 mL |
Each for $175.82
|
|
NB100165
|
Novus Biologicals
NB100165 |
0.1 mL |
Each for $436.00
|
|
Description
XRCC3 Polyclonal specifically detects XRCC3 in Human, Drosophila, Hamster samples. It is validated for Western Blot, Immunoprecipitation, ICC/IF (Negative).Specifications
XRCC3 | |
Western Blot 1:5000, Immunoprecipitation, ICC/IF (Negative) | |
Polyclonal | |
Rabbit | |
Apoptosis, Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Homologous Recombination, Tumor Suppressors | |
PBS with 0.05% Sodium Azide | |
CMM6, DNA repair protein XRCC3, X-ray repair complementing defective repair in Chinese hamster cells 3, X-ray repair cross-complementing protein 3 | |
XRCC3 | |
IgG | |
Affinity Purified | |
38 kDa |
1 mg/mL | |
Western Blot, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
RUO | |
Human, Drosophila, Hamster | |
O43542 | |
7517 | |
A synthetic partial peptide of human XRCC3. [UniProt# O43542] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title