Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

XRCC4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522044

Catalog No. PIPA522044



Description

Description

PA5-22044 targets XRCC4 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-22044 immunogen is a synthetic peptide corresponding to a region within amino acids 216 and 309 of XRCC4 (Uniprot IDNo.Q13426). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

XRCC4
Polyclonal
Unconjugated
XRCC4
Liquid
XRCC4
Synthetic peptide corresponding to a region within amino acids 216 and 309 of XRCC4 (Uniprot IDNo.Q13426)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q13426
DNA repair protein XRCC4, X-ray repair cross-complementing protein 4, X-ray repair, complementing defective, repair in Chinese hamster
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7518
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit