Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XTP4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131180

Catalog No. PIPA131180


Add to cart

Description

Description

PA1-31180 detects XTP4 from human samples. PA1-31180 has been successfully used in Western blot applications. The PA1-31180 immunogen is: Synthetic peptide: SNGETLEKITNSRPPC, corresponding to C terminal amino acids 97-112 of Human XTP4.

Specifications

Specifications

XTP4
Polyclonal
Unconjugated
MIEN1
Liquid
MIEN1
synthetic peptide SNGETLEKITNSRPPC, corresponding to C terminal amino acids 97-112 of Human XTP4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
0.5 mg/mL
Antibody with no preservative
Q9BRT3
C35, MGC14832, ORB3, RDX12, XTP4
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
84299
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit