Learn More
anti-XYLT2, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP133208
Catalog No. NBP133208
Add to cart
Description
Description
XYLT2 Polyclonal antibody specifically detects XYLT2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
XYLT2
1.0mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
XYLT2
Recombinant protein encompassing a sequence within the center region of human Xylosyltransferase II. The exact sequence is proprietary.
97 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
EC 2.4.2.26, Peptide O-xylosyltransferase 1, PXYLT2, XT2UDP-D-xylose:proteoglycan core protein beta-D-xylosyltransferase, xylosyltransferase 2, xylosyltransferase IIXT-IIprotein xylosyltransferase 2, xylT-II
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
64132
Human