Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-YB1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP197572
Description
YB1 Polyclonal specifically detects YB1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, In vivo assay, Knockdown Validated.Specifications
YB1 | |
Polyclonal | |
Western Blot 1:5000-1:10000, Simple Western 1:500, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:50-1:200, In vivo assay | |
P67809, P62960, P62961 | |
YBX1 | |
A C-terminal peptide of human YB1. [Swiss-Prot# P67809] | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse and Rat. Based on immunogen's sequence homology, this antibody is predicted to react with Bovine (100%), Rabbit (100%), Chicken (93%) and Xenopus (87%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Whole antisera with 0.05% Sodium Azide | |
BP-8, CBF-A, class II, Y box-binding protein I, CSDA2, CSDB, DBPB CCAAT-binding transcription factor I subunit A, EFI-A, Enhancer factor I subunit A, MDR-NF1, MGC104858, MGC110976, MGC117250, NSEP1 Y-box-binding protein 1, nuclease-sensitive element-binding protein 1, Y box binding protein 1, YB1 DNA-binding protein B, YB-1 Y-box transcription factor, YBX1 | |
Rabbit | |
50 kDa | |
0.1 mL | |
Breast Cancer, Cancer, DNA Repair, Epigenetics, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Transcription Factors and Regulators, Translation Control | |
4904 | |
Human, Mouse, Rat, Rabbit | |
Antisera |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction