Learn More
anti-YB1, Polyclonal, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 17250002
 View more versions of this product
Catalog No. 17-250-002
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
YB1
Unlabeled
Q15905
Rabbit
0.05mg
Primary
4904
Human
ELISA,Western Blot
20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
Affinity Purified
This antibody was made against a protein fragment from the entire range of the protein (CAA65446).
RUO
Polyclonal
This product is specific for Human YB-1.