Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YDJC Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524137
Description
Specifications
YDJC | |
Polyclonal | |
YDJC | |
UPF0249 protein ydjC homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150223, 69101 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
A8MPS7, Q14BV6 | |
YDJC | |
KLH conjugated synthetic peptide between 169-198 amino acids from the central region of human YDJC | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction