Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YES1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514800

Catalog No. PIPA514800

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

YES is a member of the Src family of non-receptor tyrosine kinases participating in a wide range of cellular signalling pathways. The gene that encodes YES is the cellular homolog of the Yamaguchi sarcoma virus oncogene. This gene lies in proximity to thymidylate synthase gene on chromosome 18, and a pseudogene of it has mapped to chromosome 22.
Specifications

Specifications

YES1
Polyclonal
PBS with 0.09% sodium azide
P07947, Q04736
Proto-oncogene tyrosine-protein kinase YES, p61-YES, C-YES
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22612, 7525
Immunohistochemistry, Western Blot
Unconjugated
YES1
Liquid
YES1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human YES1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit