Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YIPF5 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526560

Catalog No. PIPA526560

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with non-human primate based on sequence homology.

Plays a role in transport between endoplasmic reticulum and Golgi.
Specifications

Specifications

YIPF5
Polyclonal
PBS with 0.09% sodium azide
Q969M3, Q9EQQ2
FinGER5, SB140, SMAP-5, SMAP5, YIP1A, YIP1 family member 5, YPT-interacting protein 1 A, five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5, golgi membrane protein SB140, protein YIPF5, smooth muscle cell associated protein 5, smooth muscle cell-associ
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100755594, 67180, 81555
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
YIPF5
Liquid
YIPF5
KLH conjugated synthetic peptide between 21-49 amino acids from the N-terminal region of human YIPF5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit