Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF385B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525690
Description
ZNF385B contains 4 U1-type zinc fingers. ZNF385B is expressed in brain with a potential role in RNA maturation, stability, or transcription regulation. There are 5 known isoforms.Specifications
ZNF385B | |
Polyclonal | |
ZNF385B | |
ZFN533, ZNF533, zinc finger protein 533 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151126, 241494 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q569K4, Q8BXJ8 | |
Zfp385b, ZNF385B | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human Z385B | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction