Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZACN Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525013

Catalog No. PIPA525013

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

ZACN is a zinc-activated ligand-gated ion channel that defines a new subgroup of the cysteine-loop superfamily of ligand-gated ion channels.
Specifications

Specifications

ZACN
Polyclonal
PBS with 0.09% sodium azide
Q401N2
L2, LGICZ, LGICZ1, ZAC, ZAC1, ligand-gated ion channel subunit, ligand-gated ion channel zinc-activated 1, ligand-gated ion channel, zinc activated 1, ligand-gated ion-channel receptor L2, zinc activated ligand-gated ion channel 1, zinc-activated ligand-gated ion channel
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
353174
Immunohistochemistry, Western Blot
Unconjugated
ZACN
Liquid
ZACN
KLH conjugated synthetic peptide between 331-360 amino acids from the C-terminal region of human ZACN
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit