Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP-70 Kinase Mouse, Unlabeled, Clone: 29, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610239

 View more versions of this product

Catalog No. BDB610239


Add to cart

Description

Description

ZAP-70 is a protein tyrosine kinase (PTK) that associates with the ζ subunit of the T cell antigen receptor (TCR) and undergoes tyrosine phosphorylation following TCR stimulation. ZAP-70 contains two SH2-like domains with the PTK domain being at the C-terminus. It appears that both ZAP-70 and Syk are recruited to the phosphorylated CD3 and ζ subunits after TCR stimulation. The src-family PTKs, lck, and fyn, seem to play a role upstream of ZAP-70 and Syk where they phosphorylate CD3 and ζ. Thus both Src- and ZAP-70/Syk-family PTKs are critical for efficient induction of tyrosine phosphoproteins in heterologous cells. The significance of ZAP-70 in mediating TCR signal transduction has been confirmed by showing that ZAP-70 activity is absent in an autosomal recessive form of severe combined immunodeficiency (SCID). This is due to mutations affecting the ZAP-70 kinase domain which affect the stability of the protein. The levels of lck and fyn appear to be normal in the same cells. Results of the ZAP-70 mutations in these patients include an unusual absence of peripheral CD8+ T cells and an abundance of CD4+ T cells in peripheral lymphoid organs. These results indicate that these two populations of T cells develop through utilization of different intracellular signaling pathways.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

ZAP-70 Kinase
29
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA
50μg
Cell Biology
Primary
Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.