Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger, BED-type containing 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant ZBED1.

Manufacturer:  Abnova Corporation H00009189A01

Catalog No. 89-004-426


Add to cart

Description

Description

Sequence: NKSLESSQTDLKLVAHPRAKSKVWKYFGFDTNAEGCILQWKKIYCRICMAQIAYSGNTSNLSYHLEKNHPEEFCEFVKSNTEQMREAFATAFSKLKP
Specifications

Specifications

zinc finger, BED-type containing 1
Unconjugated
50% glycerol
NM_004729
ALTE|DREF|KIAA0785|TRAMP|hDREF
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
9189
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant ZBED1.
ZBED1
Serum
ZBED1
ZBED1 (NP_004720, 3 a.a. ~ 99 a.a) partial recombinant protein with GST tag.
RUO
Y
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit