Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBED3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520733

Catalog No. PIPA520733


Add to cart

Description

Description

Despite its predicted molecular weight, ZBED3 often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is Jurkat cell lysate. PA5-20733 can be used with blocking peptide PEP-0847.

Axin is a key modulator of the Wnt/beta-catenin pathway and acts as a scaffold protein in phosphorylating and degrading cytoplasmic beta-catenin. ZBED3, a member of the Zinc finger BED-type domain containing superfamily, has been recently identified as an Axin-interacting protein and has been suggested to modulate the Wnt/beta-catenin signaling pathway. RNA interference (RNAi) experiments targeting ZBED3 RNA attenuated Wnt-induced beta-catenin accumulation, lymphoid enhancer binding factor-1-dependent luciferase reporter activity, and Wnt gene expression.
Specifications

Specifications

ZBED3
Polyclonal
Unconjugated
ZBED3
Liquid
ZBED3
A 15 amino acid peptide from near the carboxy terminus of human ZBED3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q96IU2
2610005H11Rik; zinc finger BED domain-containing protein 3; zinc finger, BED domain containing 3
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
84327
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit