Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524073

Catalog No. PIPA524073


Add to cart

Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZBTB10
Polyclonal
PBS with 0.09% sodium azide
Q96DT7
RINZF, zinc finger and BTB domain-containing protein 10, zinc finger protein RIN ZF, zinc finger protein RINZF
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65986
Flow Cytometry, Western Blot
Unconjugated
ZBTB10
Liquid
ZBTB10
KLH conjugated synthetic peptide between 553-582 amino acids from the C-terminal region of human ZBTB10
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit