Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513708

Catalog No. PIPA513708

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ZBTB2 may be involved in transcriptional regulation.
Specifications

Specifications

ZBTB2
Polyclonal
PBS with 0.09% sodium azide
Q8N680
Zinc finger and BTB domain-containing protein 2; ZNF437; KIAA1483; ZBTB2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57621
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
ZBTB2
Liquid
ZBTB2
KLH conjugated synthetic peptide between 335-364 amino acids from the C-terminal region of human ZBTB2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit