Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ZBTB5, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232100

Catalog No. NBP232100


Add to cart

Description

Description

ZBTB5 Polyclonal antibody specifically detects ZBTB5 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

ZBTB5
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 mcg/mg lysate
NP_055687.1
KIAA0354, zinc finger and BTB domain containing 5, zinc finger and BTB domain-containing protein 5
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
ZBTB5
The immunogen this antibody was made to, maps to a region between residue 275 to 325 of human Zinc Finger and BTB Domain Containing 5 using the numbering given in entry NP_055687.1 (GeneID 9925).
Immunogen affinity purified
RUO
Primary
9925
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only