Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pokemon Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521087

Catalog No. PIPA521087


Add to cart

Description

Description

A suggested positive control is human ovary tissue lysate. PA5-21087 can be used with blocking peptide PEP-1201.

The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB7A, also known as LRF, was initially identified as a potential target of the LAZ-3/BCL-6 oncogene. Later reports demonstrated that it could bind to the wild-type IST (inducer of short transcripts) elements of HIV-1 long terminal repeats (LTRs) and stimulate Tat activity on the HIV-1 LTR. More recently it ZBTB7A has been recognized as a proto-oncogene whose overexpression contributes to maligcy in breast cancer and functions as a co-repressor of the androgen receptor in prostate cancer cells.
Specifications

Specifications

Pokemon
Polyclonal
Unconjugated
ZBTB7A
Liquid
ZBTB7A
A 16 amino acid peptide near the amino terminus of human ZBTB7A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O95365, O88939
FBI-1, FBI1, LRF, ZBTB7, ZNF857A, pokemon; HIV-1 1st-binding protein 1; HIV-1 inducer of short transcripts binding protein; POK erythroid myeloid ontogenic factor; POZ and Krueppel erythroid myeloid ontogenic factor; TIP21; TTF-I-interacting peptide 21; factor binding IST protein 1; factor that binds to inducer of short transcripts protein 1; leukemia/lymphoma-related factor; lymphoma related factor; zinc finger and BTB domain containing 7A, HIV-1 inducer of short transcripts binding protein; zi
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
16969, 51341
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit