Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZC3H15 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524351

Catalog No. PIPA524351

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Protects DRG1 from proteolytic degradation.
Specifications

Specifications

ZC3H15
Polyclonal
PBS with 0.09% sodium azide
Q8WU90
HT010, LEREPO4, MSTP012, DRG family-regulatory protein 1, likely ortholog of mouse immediate early response erythropoietin 4, zinc finger CCCH domain-containing protein 15
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55854
Immunohistochemistry, Western Blot
Unconjugated
ZC3H15
Liquid
ZC3H15
KLH conjugated synthetic peptide between 367-395 amino acids from the C-terminal region of human ZC3H15
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit