Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
zinc finger, C3HC-type containing 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ZC3HC1 protein.
Supplier: Abnova Corporation H00051530B01
Description
Sequence: MTDLDASFGLTSSPIPGLEGRPERLPLVPESPRRMMTRSQDATFSPGSEQAEKSPGPIVSRTRSWDSSSPVDRPEPEAASPTTRTRPVTRSMGTGDTPGLEVPSSPLRKAKRARLCSSSSSDTSSRSFFDPTSQHRDWCPWVNITLGKESRENGGTEPDASAPAEPGWKAVLTILLAHKQSSQPAETDSMSLSEKSRKVFRIFRQWESLCSCSpecifications
zinc finger, C3HC-type containing 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ZC3HC1 protein. | |
ZC3HC1 | |
1110054L24Rik/HSPC216/NIPA | |
Mouse | |
50 μL | |
Yes | |
51530 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC028917.1 | |
ZC3HC1 | |
ZC3HC1 (AAH28917.1, 1 a.a. ∼ 212 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'