Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523820

Catalog No. PIPA523820

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ZDHHC1
Polyclonal
PBS with 0.09% sodium azide
Q8WTX9, Q8R0N9
C16orf1, HSU90653, ZNF377, DHHC domain-containing cysteine-rich protein 1, DHHC-1, DHHC-domain-containing cysteine-rich protein, probable palmitoyltransferase ZDHHC1, zinc finger DHHC domain-containing protein 1, zinc finger protein 377, zinc finger, DHHC domain containing 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29800, 70796
Western Blot
Unconjugated
ZDHHC1
Liquid
ZDHHC1
KLH conjugated synthetic peptide between 14-43 amino acids from the N-terminal region of human ZDHHC1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit