Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZDHHC7, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ZDHHC7 protein.
Supplier: Abnova Corporation H00055625B01P
Description
Sequence: MLTDPGAVPKGNATKEYMESLQLKPGEVIYKCPKCCCIKPERAHHCSICKRCIRKMDHHCPWVNNCVGEKNQRFFVLFTMYIALSSVHALILCGFQFISCVRGQWTECSDFSPPITVILLIFLCLEGLLFFTFTAVMFGTQIHSICNDETEIERLKSEKPTWERRLRWEGMKSVFGGPPSLLWMNPFVGFRFRRLPTRPRKGGPEFSVSpecifications
ZDHHC7 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ZDHHC7 protein. | |
ZDHHC7 | |
FLJ10792/FLJ20279/ZNF370 | |
Mouse | |
Affinity chromatography | |
RUO | |
55625 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC017702 | |
ZDHHC7 | |
ZDHHC7 (AAH17702, 1 a.a. ∼ 208 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction