Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC9 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526721

Catalog No. PIPA526721


Add to cart

Description

Description

This antibody is predicted to react with bovine and non-human primate based on sequence homology.

The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.
Specifications

Specifications

ZDHHC9
Polyclonal
PBS with 0.09% sodium azide
Q9Y397, P59268
CGI89, CXorf11, DHHC9, MMSA1, MRXSZ, ZDHHC10, ZNF379, ZNF380, Asp-His-His-Cys domain containing protein 9, antigen MMSA-1, palmitoyltransferase ZDHHC9, zinc finger protein 379, zinc finger protein 380, zinc finger, DHHC domain containing 10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
208884, 51114
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ZDHHC9
Liquid
ZDHHC9
KLH conjugated synthetic peptide between 335-364 amino acids from the C-terminal region of human ZDHHC9
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit