Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZEB1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP213159
Description
ZEB1 Polyclonal specifically detects ZEB1 in Human, Primate samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.Specifications
ZEB1 | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.01% Sodium Azide | |
AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Primate | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1.00 mg/mL | |
Western Blot 1:1000, Simple Western 1:200, Immunocytochemistry/Immunofluorescence 1:40 | |
P37275 | |
ZEB1 | |
A synthetic peptide made to an C-terminal portion of the human ZEB1 protein (between residues 1087-1124). [Uniprot: P37275] | |
0.1 mL | |
Cancer, Immunology, Neuroscience, Transcription Factors and Regulators, Vision, Zinc Finger | |
6935 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction