Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFYVE21 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA534408
Description
A suggested positive control is NIH-3T3 cell lysate. PA5-34408 can be used with blocking peptide PEP-1450.
ZFYVE21 Polyclonal antibody specifically detects ZFYVE21 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ZFYVE21 | |
Polyclonal | |
Unconjugated | |
ZFYVE21 | |
1110013H04Rik; AW558780; C85416; FYVE domain containing 21; HCV p7-transactivated protein 1; HCVP7TP1; ZF21; Zfyve21; Zinc finger FYVE domain-containing protein 21; zinc finger FYVE-type containing 21; zinc finger, FYVE domain containing 21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
362789, 68520, 79038 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
D3ZVP7, Q8VCM3, Q9BQ24 | |
ZFYVE21 | |
A 15 amino acid synthetic peptide near the amino terminus of human ZF21 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction