Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIC3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA529073

Catalog No. PIPA529073


Add to cart

Description

Description

PA5-29073 targets ZIC3 in IF and WB applications and shows reactivity with Human samples. The PA5-29073 immunogen is recombit fragment corresponding to a region within amino acids 172 and 460 of Human ZIC3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs.
Specifications

Specifications

ZIC3
Polyclonal
Unconjugated
ZIC3
Liquid
ZIC3
Recombit fragment corresponding to a region within amino acids 172 and 460 of Human ZIC3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
0.84 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O60481
HTX, HTX1, ZNF203, Zic family member 3 (odd-paired homolog, Drosophila), heterotaxy 1, zinc finger protein 203, zinc finger protein ZIC 3, zinc finger protein of the cerebellum 3
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7547
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit