Learn More
ZIC3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524604
Catalog No. PIPA524604
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs.
Specifications
Specifications
ZIC3
Polyclonal
Unconjugated
ZIC3
Liquid
ZIC3
KLH conjugated synthetic peptide between 239-267 amino acids from the central region of human ZIC3
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide
O60481
HTX, HTX1, ZNF203, Zic family member 3 (odd-paired homolog, Drosophila), heterotaxy 1, zinc finger protein 203, zinc finger protein ZIC 3, zinc finger protein of the cerebellum 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7547