Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIK1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524079

Catalog No. PIPA524079

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be a transcriptional repressor.
Specifications

Specifications

ZIK1
Polyclonal
PBS with 0.09% sodium azide
Q3SY52
ZNF762, zinc finger protein 762, zinc finger protein interacting with ribonucleoprotein K
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
284307
Flow Cytometry, Western Blot
Unconjugated
ZIK1
Liquid
ZIK1
KLH conjugated synthetic peptide between 186-215 amino acids from the central region of human ZIK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit