Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIPK Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA130693

Catalog No. PIPA130693

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-30693 detects ZIP Kinase from mouse, rat, human samples. PA1-30693 has been successfully used in Western blot applications. The PA1-30693 immunogen is: synthetic peptide corresponding to amino acids 279 to 298 of human origin. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Cell death signals are transduced by death domain containing adaptor molecules and members of the caspase family of proteases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZIPK
Polyclonal
Unconjugated
O43293, O54784, O88764
DAPK3
synthetic peptide corresponding to amino acids 279 to 298 of human origin.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Western Blot
0.5 mg/mL
DAPK3
Death-associated protein kinase 3, ZIPK, DAP kinase 3, Dlk
Rabbit
Antigen affinity chromatography
RUO
13144, 1613, 64391
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit