Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIPK Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112521

Catalog No. 01-674-181


Add to cart

Description

Description

A suggested positive control for this product is Jurkat or HeLa whole cell lysate.

Apoptosis is mediated by death domain containing adapter molecules and a caspase family of proteases. Certain serine/threonine protein kinases, such as ASK-1 and RIP, are mediators of apoptosis. Two novel serine/threonine kinases that induce apoptosis were recently identified and designated DRAK1 and DRAK2 for DAP kinase-related apoptosis-inducing protein kinases1. DRAKs contain an N-terminal kinase domain and a C-terminal regulation domain. Overexpression of DRAK1 induces apoptosis. DRAKs have high sequence homology to DAP and ZIP kinases, and they represent a novel family of serine/threonine kinases, which mediates apoptosis through their catalytic activities. DRAK1 is located in nucleus and the messenger RNA was ubiquitously expressed in human tissues.
Specifications

Specifications

ZIPK
Polyclonal
PBS with 0.02% sodium azide
O43293, O54784, O88764
DAP3K, ZIP
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
13144, 1613, 64391
Western Blot
Unconjugated
DAPK3
Liquid
DAPK3
Synthetic peptide corresponding to residues 279-298 of human ZIP kinase.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit