Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC39A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521067
Description
A suggested positive control is rat brain tissue lysate. PA5-21067 can be used with blocking peptide PEP-1181.
SLC39A2 Polyclonal antibody specifically detects SLC39A2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SLC39A2 | |
Polyclonal | |
Unconjugated | |
SLC39A2 | |
6A1; ETI-1; F730005G13Rik; Gm1789; Gm289; MGC119190; SLC39A2; solute carrier family 39 (zinc transporter), member 2; solute carrier family 39 member 2; Zinc transporter ZIP2; zinc uptake transporter 2; ZIP2; ZIP-2; Zrt- and Irt-like protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
214922, 29986, 305846 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9NP94 | |
SLC39A2 | |
A 17 amino acid peptide near the amino terminus of human ZIP2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction