Learn More
TEX11, clone: 236734, Invitrogen™
Rat Monoclonal Antibody
Manufacturer:  Invitrogen MA523944
Catalog No. PIMA523944
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
TEX11
Monoclonal
0.5mg/mL
PBS with 5% trehalose and no preservative
Q14AT2
Tex11, TGC1, TSGA3, testis expressed 11, testis expressed sequence 11, testis-expressed sequence 11 protein, TGC1, TSGA3
Rat
IgG2B
100μg
Store at-20°C, Avoid Freeze/Thaw Cycles
Primary
83558
Western Blot
236734
Unconjugated
Tex11
Lyophilized
TEX11
E. coli-derived recombinant mouse ZIP4H/Tex11 Met768-Leu947
Protein A/G
RUO
Antibody
Monoclonal
Murine