Learn More
ZMPSTE24 Rabbit anti-Human, Mouse, Primates, Zebra fish, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of ZMPSTE24.
Manufacturer: Abnova Corporation PAB11975
Description
This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy. [provided by RefSeq
Specifications
ZMPSTE24 | |
Unconjugated | |
Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
ZMPSTE24 | |
FACE-1|FACE1|FLJ14968|STE24|Ste24p | |
Rabbit | |
100 uL | |
Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
This antibody is specific to ZMPSTE24. |
Western Blot | |
Rabbit polyclonal antibody raised against synthetic peptide of ZMPSTE24. | |
In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
Liquid | |
ZMPSTE24 | |
A synthetic peptide corresponding to amino acids 1-100 of human ZMPSTE24. | |
RUO | |
Primary | |
10269 | |
Human, Murine, Primate, Zebrafish |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only