Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ZMPSTE24, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $314.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide made to a C-terminal portion of the human ZMPSTE24 protein sequence (between residues 400-475). [Swiss-Prot# O75844]
Antigen ZMPSTE24
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1002388SS
View Documents
Novus Biologicals
NB1002388SS
0.025 ml
Each for $119.60
Add to cart
 
NB1002388
View Documents
Novus Biologicals
NB1002388
0.1 ml
Each for $314.00
Add to cart
 
Description

Description

ZMPSTE24 Polyclonal antibody specifically detects ZMPSTE24 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Rabbit
A synthetic peptide made to a C-terminal portion of the human ZMPSTE24 protein sequence (between residues 400-475). [Swiss-Prot# O75844]
Polyclonal
Primary
Western Blot
10269
1.0 mg/mL
IgG
ZMPSTE24
Affinity Purified
RUO
O75844
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit