Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZMYM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521089

Catalog No. PIPA521089


Add to cart

Description

Description

A suggested positive control is rat lung tissue lysate. PA5-21089 can be used with blocking peptide PEP-1203.

Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. As a member of the MYM (myeloproliferative and mental retardation) gene family, ZMYM1 is widely expressed in different tissues in eukaryotes under several forms derived by alternative splicing. While its function remains unknown, the related protein ZMYM2 has been shown to associate with and stabilize the LSD1-CoREST-HDAC1 (LCH) complex of chromatin through its MYM-type zinc fingers, thereby enhancing the transcriptional repression of several genes, suggesting that ZMYM1 may play a similar role.
Specifications

Specifications

ZMYM1
Polyclonal
Unconjugated
ZMYM1
Liquid
ZMYM1
A 17 amino acid peptide near the carboxy terminus of human ZMYM1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q5SVZ6
RP11-248I9.4, MYM, RP11-181E22.4; zinc finger MYM-type protein 1; zinc finger, MYM domain containing 1
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
313604, 68310, 79830
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit