Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZMYM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521090

Catalog No. PIPA521090


Add to cart

Description

Description

A suggested positive control is rat lung tissue lysate. PA5-21090 can be used with blocking peptide PEP-1204. The immunogen is located within amino acids 120 - 170 of ZMYM2.

Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. As a member of the MYM (myeloproliferative and mental retardation) gene family, ZMYM1 is widely expressed in different tissues in eukaryotes under several forms derived by alternative splicing. While its function remains unknown, the related protein ZMYM2 has been shown to associate with and stabilize the LSD1-CoREST-HDAC1 (LCH) complex of chromatin through its MYM-type zinc fingers, thereby enhancing the transcriptional repression of several genes, suggesting that ZMYM1 may play a similar role.
Specifications

Specifications

ZMYM2
Polyclonal
Unconjugated
ZMYM2
Liquid
ZMYM2
A 17 amino acid peptide near the amino terminus of human ZMYM2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9UBW7, Q9CU65
RP11-264J4.7, FIM, MYM, RAMP, SCLL, ZNF198; fused in myeloproliferative disorders protein; rearranged in an atypical myeloproliferative disorder; zinc finger MYM-type protein 2; zinc finger protein 198
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
305913, 76007, 7750
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit