Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZMYM3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521091
Description
A suggested positive control is human brain tissue lysate. PA5-21091 can be used with blocking peptide PEP-1205.
ZMYM3 Polyclonal antibody specifically detects ZMYM3 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ZMYM3 | |
Polyclonal | |
Unconjugated | |
ZMYM3 | |
9030216B10Rik; AW122925; DXHXS6673E protein; DXS6673E; DXS6673El; KIAA0385; MYM; XFIM; Zfp261; zinc finger MYM-type containing 3; zinc finger MYM-type protein 3; Zinc finger protein 261; zinc finger, MYM-type 3; Zmym3; ZNF198L2; ZNF261 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56364, 9203 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q14202, Q9JLM4 | |
ZMYM3 | |
The immunogen is located within amino acids 220 - 270 of human ZMYM3. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction