Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZMYND10 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525732

Catalog No. PIPA525732


Add to cart

Description

Description

This antibody is predicted to react with rat based on sequence homology.

Specifications

Specifications

ZMYND10
Polyclonal
PBS with 0.09% sodium azide
O75800, Q99ML0
BLU, FLU, protein BLu, zinc finger MYND domain-containing protein 10, zinc finger, MYND domain containing 10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
114602, 51364
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
ZMYND10
Liquid
ZMYND10
KLH conjugated synthetic peptide between 404-429 amino acids from the C-terminal region of human ZMYND10
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit