Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF160 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526907
Description
The protein encoded by this gene is a Kruppel-related zinc finger protein which is characterized by the presence of an N-terminal repressor domain, the Kruppel-associated box (KRAB). The KRAB domain is a potent repressor of transcription; thus this protein may function in transcription regulation.Specifications
ZNF160 | |
Polyclonal | |
ZNF160 | |
F11, HKr18, HZF5, KR18, KRAB zinc finger protein KR18, zinc finger protein 5, zinc finger protein HZF5, zinc finger protein Kr18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90338 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q9HCG1 | |
ZNF160 | |
KLH conjugated synthetic peptide between 143-171 amino acids from the N-terminal region of human ZN160 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction