Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 133 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant ZNF133.

Manufacturer:  Abnova Corporation H00007692A01

Catalog No. 89-004-369


Add to cart

Description

Description

Sequence: PGPGDQEKQQQASEGRPWSDQAEGPEGEGAMPLFGRTKKRTLGAFSRPPQRQPVSSRNGLRGVELEASPAQTGNPEETDKLLKRIEVLGFGTVNCGECGLSFSKMTNLLS
Specifications

Specifications

zinc finger protein 133
Unconjugated
50% glycerol
NM_003434
ZNF150|pHZ-13|pHZ-66
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant ZNF133.
ZNF133
Serum
ZNF133
ZNF133 (NP_003425, 121 a.a. ~ 230 a.a) partial recombinant protein with GST tag.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7692
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit