Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 157, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ZNF157 protein.

Manufacturer:  Abnova Corporation H00007712B01

Catalog No. 89-006-495


Add to cart

Description

Description

This gene product is a likely zinc finger family transcription factor. It contains KRAB-A and KRAB-B domains that act as transcriptional repressors in related proteins, and multiple zinc finger DNA binding motifs and finger linking regions characteristic of the Kruppel family. This gene is part of a gene cluster on chromosome Xp11.23. [provided by RefSeq

Sequence: MPANGTSPQRFPALIPGEPGRSFEGSVSFEDVAVDFTRQEWHRLDPAQRTMHKDVMLETYSNLASVGLCVAKPEMIFKLERGEELWILEEESSGHGYSGSLSLLCGNGSVGDNALRHDNDLLHHQKIQTLDQNVEYNGCRKAFHEKTGFVRRKRTPRGDKNFECHECGKAYCRKSNLVEHLRIHTGERPYECGECAKTFSARSYLIAHQKTHTGERPFECNECGKSFGRKSQLILHTRTHTGERPYECTECGKTFSEKATLTIHQRTHTGEKPYECSECGKTFRVKISLTQHHRTHTGEKPYECGECGKNFRAKKSLNQHQRIHTGEKPYECGECGKFFRMKMTLNNHQRTHTGEKPYQCNECGKSFRVHSSLGIHQRIHTGEKPYECNECGNAFYVKARLIEHQRMHSGEKPYECSECGKIFSMKKSLCQHRRTHTGEKPYECSECGNAFYVKVRLIEHQRIHTGERPFECQECGKAFCRKAHLTEHQRTHIGWSWRCTMKKASH
Specifications

Specifications

zinc finger protein 157
Unconjugated
No additive
NM_003446.3
HZF22
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human ZNF157 protein.
ZNF157
Serum
ZNF157
ZNF157 (NP_003437.2, 1 a.a. ~ 506 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7712
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit