Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 165, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ZNF165 protein.

Manufacturer:  Abnova Corporation H00007718B01

Catalog No. 89-006-496


Add to cart

Description

Description

This gene encodes a member of the Kruppel family of zinc finger proteins. Members of this DNA-binding protein family act as transcriptional regulators. This gene is located within a cluster of zinc finger family members. The encoded protein may play a role in spermatogenesis. [provided by RefSeq

Sequence: MATEPKKAAAQNSPEDEGLLIVKIEEEEFIHGQDTCLQRSELLKQELCRQLFRQFCYQDSPGPREALSRLRELCCQWLKPEIHTKEQILELLVLEQFLTILPGDLQAWVHEHYPESGEEAVTILEDLERGTDEAVLQVQAHEHGQEIFQKKVSPPGPALNVKLQPVETKAHFDSSEPQLLWDCDNESENSRSMPKLEIFEKIESQRIISGRISGYISEASGESQDICKSAGRVKRQWEKESGESQRLSSAQDEGFGKILTHKNTVRGEIISHDGCERRLNLNSNEFTHQKSCKHGTCDQSFKWNSDFINHQIIYAGEKNHQYGKSFKSPKLAKHAAVFSGDKTHQCNECGKAFRHSSKLARHQRIHTGERCYECNECGKSFAESSDLTRHRRIHTGERPFGCKECGRAFNLNSHLIRHQRIHTREKPYECSECGKTFRVSSHLIRHFRIHTGEKPYECSECGRAFSQSSNLSQHQRIHMRENLLM
Specifications

Specifications

zinc finger protein 165
Unconjugated
No additive
NM_003447.2
LD65|ZSCAN7
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7718
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human ZNF165 protein.
ZNF165
Serum
ZNF165
ZNF165 (NP_003438.1, 1 a.a. ~ 485 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit