Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF197 Mouse anti-Human, Clone: 3C11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA520946

Catalog No. PIMA520946


Add to cart

Description

Description

Peptide Sequence: IAAEICPHPP TDLVAFNLQD PQHDSPAPEA SALSQEENPR NQLMALMLLT AQPQELVMFE EVSVCFTSEE WACLGPIQRA LYWDVMLENY GNVTSLGYRK YRRQRNK

ZNF197 product belongs to the zinc finger protein superfamily, members of which are regulatory proteins characterized by nucleic acid-binding zinc finger domains. The encoded protein contains 20 tandemly arrayed C2H2-type zinc fingers, a Kruppel-associated box (KRAB) domain, and a SCAN box. This transcript turns over rapidly and contains 3 ft. UTR AUUUA motifs, which are often a hallmark of rapid turnover. It is overexpressed in some thyroid papillary carcinomas. ZNF197 is located in a cluster of zinc finger genes at 3p21. Two alternatively spliced transcripts encoding different isoforms have been described.
Specifications

Specifications

ZNF197
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
O14709
D3S1363E, P18, VHLaK, ZKSCAN9, ZNF166, ZNF20, VHL-associated KRAB-A domain-containing protein, zinc finger protein 166, zinc finger protein 20, zinc finger protein with KRAB and SCAN domains 9
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10168
ELISA, Western Blot
3C11
Unconjugated
ZNF197
Liquid
ZNF197
ZNF197 (NP_001020026.1, 161 a.a. ∽ 267 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit