Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF197 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524033

Catalog No. PIPA524033

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ZNF197 product belongs to the zinc finger protein superfamily, members of which are regulatory proteins characterized by nucleic acid-binding zinc finger domains. The encoded protein contains 20 tandemly arrayed C2H2-type zinc fingers, a Kruppel-associated box (KRAB) domain, and a SCAN box. This transcript turns over rapidly and contains 3 ft. UTR AUUUA motifs, which are often a hallmark of rapid turnover. It is overexpressed in some thyroid papillary carcinomas. ZNF197 is located in a cluster of zinc finger genes at 3p21. Two alternatively spliced transcripts encoding different isoforms have been described.
Specifications

Specifications

ZNF197
Polyclonal
PBS with 0.09% sodium azide
O14709
D3S1363E, P18, VHLaK, ZKSCAN9, ZNF166, ZNF20, VHL-associated KRAB-A domain-containing protein, zinc finger protein 166, zinc finger protein 20, zinc finger protein with KRAB and SCAN domains 9
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10168
Immunohistochemistry, Western Blot
Unconjugated
ZNF197
Liquid
ZNF197
KLH conjugated synthetic peptide between 314-343 amino acids from the N-terminal region of human ZNF197
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit