Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF230 Goat anti-Bovine, Canine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131904

Catalog No. PIPA131904

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-31918 detects SmarcAL1 from human samples. PA1-31918 has been successfully used in Western blot applications. The PA1-31918 immunogen is: Synthetic peptide: SNADQRPHDSHSFQAKGC, corresponding to amino acids 80-96 of Human SmarcAL1.

Specifications

Specifications

ZNF230
Polyclonal
Unconjugated
RNF141
Liquid
RNF141
synthetic peptide LNMADEAGQPHRP from C Terminus of the protein sequence according to NP_057506.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Canine, Human, Mouse, Rat
ELISA, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q32L15, Q2XNS1, Q8WVD5, Q99MB7, Q6IV57
FDZF2
Goat
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
308900, 476852, 50862, 539455, 67150
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit