Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 24, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ZNF24 protein.

Manufacturer:  Abnova Corporation H00007572B01

Catalog No. 89-006-472


Add to cart

Description

Description

Sequence: MSAQSVEEDSILIIPTPDEEEKILRVKLEEDPDGEEGSSIPWNHLPDPEIFRQRFRQFGYQDSPGPREAVSQLRELCRLWLRPETHTKEQILELVVLEQFVAILPKELQTWVRDHHPENGEEAVTVLEDLESELDDPGQPVSLRRRKREVLVEDMVSQEEAQGLPSSELDAVENQLKWASWELHSLRHCDDDGRTENGALAPKQELPSALESHEVPGTLSMGVPQIFKYGETCFPKGRFERKRNPSRKKQHICDECGKHFSQGSALILHQRIHSGEKPYGCVECGKAFSRSSILVQHQRVHTGEKPYKCLECGKAFSQNSGLINHQRIHTGEKPYECVQCGKSYSQSSNLFRHQRRHNAEKLLNVVKV
Specifications

Specifications

zinc finger protein 24
Unconjugated
No additive
BC003566.1
KOX17|RSG-A|ZNF191|ZSCAN3|Zfp191
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human ZNF24 protein.
ZNF24
Serum
ZNF24
ZNF24 (AAH03566.1, 1 a.a. ~ 368 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7572
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit